Human Recombinant IL-7 R alpha & TSLP R (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Human IL-7 RA&TSLP R Heterodimer Protein, Fc Tag&Fc Tag (Cat. No. ILR-H5255) on Protein A Biosensor, can bind Human TSLP, His Tag (Cat. No. TSP-H52Hb) with an affinity constant of 134 pM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P16871-1 (IL-7RA) & Q9HC73-1 (TSLPR);3575 | 64109;NP_002176.2 | NP_071431.2
- Protein Synonyms:IL7Ra,CD127,TSLP R,CRLF2,IL-XR,ILXR,p2RY8,CRLF2 fusion,TSLP receptor,TSLPR,CRL2,CRLF2Y,Cytokine receptor-like 2
- Protein/Peptide Name:IL-7 R alpha & TSLP R
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:51.7 kDa (IL-7 RA) & 50.3 kDa (TSLP R)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together