SARS-CoV-2 Recombinant S1 protein NTD (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:SARS-CoV-2
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:QHD43416.1 (P272L)
- Protein Synonyms:S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD
- Protein/Peptide Name:S1 protein NTD
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:34.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together