Human Recombinant IL-1 Rrp2 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q9HB29-1 (C154S, C262S);8808;NP_003845.2
- Protein Synonyms:IL-1 R6,IL-1 Rrp2,IL1RL2,IL-1RL2,IL1Rrp2,IL-1Rrp2,IL1RRP2,IL-36R
- Protein/Peptide Name:IL-1 Rrp2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:37.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together