Human Recombinant IL-2 R beta & IL-2 R gamma (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P14784-1 (IL-2RB) & P31785-1 (IL-2RG);3560 | 3561;NP_000869.1 (IL-2RB) & AAH14972.1 (IL-2RG)
- Protein Synonyms:IL-2 R beta & IL-2 R gamma,IL-2RB & IL-2RG
- Protein/Peptide Name:IL-2 R beta & IL-2 R gamma
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:52.8 kDa | 55.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together