Human Recombinant CCR8 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–70℃
- Biological Activity:Please contact us for detailed information.
- Gene ID:P51685-1
- Protein Synonyms:CC chemokine receptor 8,CD198,,ChemR1,CKRL1,CKR-L1,CKRL1,CY6,GPRCL1,GPRCY6,GPR-CY6,TER-1
- Protein/Peptide Name:CCR8
- Molecular Weight:43.4 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:Dry Ice
Specifications
Frequently Bought Together