Canine Recombinant IL-3 R alpha (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:609293;XP_038305195.1
- Protein Synonyms:IL3R,IL3RA,IL-3Ra,IL-3R-alpha,IL3RAY,IL3RX,IL3RY,CD123 antigen,CD123,hIL3Ra,hIL-3Ra,MGC34174,IL-3 R alpha
- Protein/Peptide Name:IL-3 R alpha
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:34.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together