Rabbit Recombinant CD3E & CD3D (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Rabbit
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q9TUF9-1 (CD3E) & A0A5F9D3I4-1 (CD3D)
- Protein Synonyms:CD3E & CD3D,CD3 delta & CD3 epsilon,CD3 delta , epsilon
- Protein/Peptide Name:CD3E & CD3D
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:16.5 kDa | 14.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together