SARS-CoV-2 Recombinant S protein RBD (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:SARS-CoV-2
- Storage Conditions:–70℃
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Supplied as 0.2 μm filtered solution in PBS, pH7.4.
- Gene ID:QHD43416.1
- Protein Synonyms:Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD
- Protein/Peptide Name:S protein RBD
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:67.1 kDa
- Endotoxin Level:Low
- Formulation:Supplied as 0.2 μm filtered solution in PBS, pH7.4.
- Shipping Temperature:Dry Ice
Specifications
Frequently Bought Together