Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Unconjugated
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    SARS-CoV-2
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    QHD43416.1 (K417T, E484K, N501Y)
  • Protein Synonyms:
    Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD
  • Protein/Peptide Name:
    S protein RBD
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    26.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together