Cynomolgus Recombinant BAFFR (from HEK293), Unconjugated, Acro Biosystems
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:A0A2K5V1K1;102116472;XP_005567184.1
- Protein Synonyms:BAFFR,TNFRSF13C,BROMIX,CD268,CVID4,prolixin,BAFF-R
- Protein/Peptide Name:BAFFR
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:9.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together