Order Entry
United States
ContactUsLinkComponent
Human Recombinant Siglec-2 (from HEK293), Unconjugated
Human Recombinant Siglec-2 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Human Recombinant Siglec-2 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P20273-1;933;NP_001762.2
  • Protein Synonyms:
    CD22,SIGLEC2,BL-CAM,SIGLEC-2,Siglec2,SIGLEC2FLJ22814
  • Protein/Peptide Name:
    Siglec-2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    101.6 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together