Rat Recombinant CD3E & CD3G (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Rat
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:315609 | 300678;NP_001101610.1 (CD3E) & NP_001071114.1 (CD3G)
- Protein Synonyms:CD3 epsilon & CD3 gamma,CD3E & CD3G
- Protein/Peptide Name:CD3E & CD3G
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:16.7 kDa | 15.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together