Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant CD79B (from HEK293), Unconjugated
Mouse Recombinant CD79B (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Mouse Recombinant CD79B (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 0.1 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, 0.2M Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P15530-1;15985;NP_032365.1
  • Protein Synonyms:
    CD79b,B29,IGB,Ig-beta
  • Protein/Peptide Name:
    CD79B
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    17.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, 0.2M Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together