Human Recombinant CD79B (from HEK293), Unconjugated
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, 0.2 M Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P40259-1;974;NP_000617.1
- Protein Synonyms:CD79b,B29,IGB,Ig-beta
- Protein/Peptide Name:CD79B
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:17.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, 0.2 M Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together