Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD74 (from HEK293), Unconjugated
Human Recombinant CD74 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Human Recombinant CD74 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 0.1 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P04233-2;972;NP_004346.1
  • Protein Synonyms:
    CD74,DHLAG,HLADG,II,Ia-GAMMA,p33,CLIP,INVG34
  • Protein/Peptide Name:
    CD74
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    47.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together