Mouse Recombinant CEACAM-5 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Mouse
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:Q3UKK2-1
- Protein Synonyms:CEACAM-5,CD66e,CEA,Meconium antigen 100
- Protein/Peptide Name:CEACAM-5
- Molecular Weight:104.5 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together