Human Recombinant Complement C5 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:P01031-1
- Protein Synonyms:Complement C5,C5,CPAMD4
- Protein/Peptide Name:Complement C5
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together