SARS-CoV-2 Recombinant S1 protein (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:SARS-CoV-2
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 0.2 M Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:QHD43416.1
- Protein Synonyms:Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1
- Protein/Peptide Name:S1 protein
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:101.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 0.2 M Arginine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together