Human Recombinant NCAM-1 (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 500 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P13591-2;4684;NP_851996.2
- Protein Synonyms:CD56,MSK39,NCAM1,N-CAM-1
- Protein/Peptide Name:NCAM-1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:81.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 500 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together