Human Recombinant TLR3 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:Q6PCD4-1
- Protein Synonyms:TLR3,CD283,IIAE2
- Protein/Peptide Name:TLR3
- Molecular Weight:103.7 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together