SARS-CoV-2 Recombinant Nucleocapsid protein (from HEK293), Unconjugated
- Conjugation:Unconjugated
- Species:SARS-CoV-2
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:QHO62115.1 (S194L)
- Protein Synonyms:Nucleocapsid protein,NP,Protein N
- Protein/Peptide Name:Nucleocapsid protein
- Molecular Weight:47.3 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Frequently Bought Together