Human Recombinant CD2 (from HEK293), Precision Avi
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Gene ID:P06729-1(H266Q);914;AAH33583
- Protein Synonyms:CD2,SRBC,LFA-2,T11
- Protein/Peptide Name:CD2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:24.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together