Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Integrin alpha 10 beta 1 (from HEK293), Unconjugated
Mouse Recombinant Integrin alpha 10 beta 1 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Mouse Recombinant Integrin alpha 10 beta 1 (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    E9PXZ3-1 (ITGA10) & P09055-1 (ITGB1);213119 | 16412;NP_001289400.1 | NP_034708.1
  • Protein Synonyms:
    Integrin alpha 10 beta 1,ITGA10&ITGB1
  • Protein/Peptide Name:
    Integrin alpha 10 beta 1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    126.6 kDa (ITGA10) & 83.5 kDa (ITGB1)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together