Mouse Recombinant Integrin alpha 11 beta 1 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P61622-1 (ITGA11) & P09055-1 (ITGB1); | 16412; | NP_034708.1
- Protein Synonyms:Integrin alpha 11 beta 1,ITGA11&ITGB1
- Protein/Peptide Name:Integrin alpha 11 beta 1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:132.0 kDa (ITGA11) & 83.5 kDa (ITGB1)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together