Human Recombinant MED1 (from InsectCells), Unconjugated
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:Q15648-1
- Protein/Peptide Name:MED1
- Molecular Weight:170.3 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together