Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant IL-2 R beta & IL-2 R gamma (from HEK293), Unconjugated
Mouse Recombinant IL-2 R beta & IL-2 R gamma (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Mouse Recombinant IL-2 R beta & IL-2 R gamma (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P16297-1 (IL-2RB) & P34902-1 (IL-2RG);16185 | 16186;NP_032394.1 | NP_038591.1
  • Protein Synonyms:
    IL-2 R beta & IL-2 R gamma,IL-2RB & IL-2RG
  • Protein/Peptide Name:
    IL-2 R beta & IL-2 R gamma
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    30.4 kDa | 35.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together