Human Recombinant LSECtin (from HEK293), Unconjugated
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:Q6UXB4-1;339390;NP_940894.1
- Protein Synonyms:CLEC4G,LSECtin
- Protein/Peptide Name:LSECtin
- Molecular Weight:28.4 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Frequently Bought Together