Mouse Recombinant CD36 (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q08857-1;12491;NP_031669.3
- Protein Synonyms:CD36,SCARB3,BDPLT10,CHDS7,FAT,GP3B,GP4,GPIV,PASIV,Platelet Glycoprotein 4,glycoprotein IV,gpIV,glycoprotein IIIb,gpIIIb
- Protein/Peptide Name:CD36
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:48.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together