Rat Recombinant IL-2 R beta & IL-2 R gamma (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Rat
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Biotinylated Human IL-2, Fc,Avitag (Cat. No. IL2-H82F3) on SA Biosensor, can bind Rat IL-2RB&IL-2RG Heterodimer Protein, His Tag&Twin-Strep Tag (Cat. No. ILG-R5283) with an affinity constant of 3.50 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P26896-1 (IL-2RB) & Q68FU6-1 (IL-2RG);25746 | 140924;NP_037327.1 | NP_543165.1
- Protein Synonyms:IL-2 R beta & IL-2 R gamma,IL-2RB & IL-2RG
- Protein/Peptide Name:IL-2 R beta & IL-2 R gamma
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:30.2 kDa | 34.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together