Human Recombinant IL-1 beta (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Gene ID:P01584-1;3553;NP_000567.1
- Protein Synonyms:IL1B,IL-1BETA,IL1F2,IL-1β
- Protein/Peptide Name:IL-1 beta
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:43.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together