Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL-3 R alpha (from HEK293), Unconjugated
Human Recombinant IL-3 R alpha (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Human Recombinant IL-3 R alpha (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Anti-IL-3 R alpha Antibody, Human IgG1 immobilized on CM5 Chip can bind Human IL-3 R alpha, Fc Tag (Cat. No. ILA-H5252) with an affinity constant of 50.3 nM as determined in a SPR assay (Biacore T200) (QC tested).
  • Gene ID:
    P26951-1;3563;NP_002174.1
  • Protein Synonyms:
    IL3R,IL3RA,IL-3Ra,IL-3R-alpha,IL3RAY,IL3RX,IL3RY,CD123 antigen,CD123,hIL3Ra,hIL-3Ra,MGC34174,IL-3 R alpha
  • Protein/Peptide Name:
    IL-3 R alpha
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    59.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together