Human Recombinant PD-L1 (from HEK293), Precision Avi
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:NP_054862.1
- Protein Synonyms:PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1
- Protein/Peptide Name:PD-L1
- Molecular Weight:53.8 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Frequently Bought Together