Human Recombinant CTLA-4 (from HEK293), Precision Avi
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:Q6GR94-1
- Protein Synonyms:CTLA4,CD152
- Protein/Peptide Name:CTLA-4
- Molecular Weight:42.1 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together