Eisco LabGlass® Reagent Bottles with Polypropylene Stopper, Amber, Narrow Mouth
Supplier: Wards
Certificates
About this item
High-quality amber glass reagent bottle useful for storing light-sensitive chemicals in classrooms or laboratories.
- Made of borosilicate glass 3.3
- Interchangeable, acid-proof polypropylene stopper
- Ungraduated
- Narrow mouth design
- Fully autoclavable and top rack dishwasher safe
- Superior chemical durability
- Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: Strain point 515 °C; annealing point 565 °C; softening point 820 °C.
Frequently Bought Together