Order Entry
United States
ContactUsLinkComponent
Ultra Flex Support System with Base Plate, Cole-Parmer
Ultra Flex Support System with Base Plate, Cole-Parmer
Catalog # MSPP-EW0802902
Supplier:  Cole Parmer
undefined
Ultra Flex Support System with Base Plate, Cole-Parmer
Catalog # MSPP-EW0802902
Supplier:  Cole Parmer
Supplier Number:  08029-02

Specifications

  • Description:
    Ultra flex support with base plate, 18" arm length
  • Dimensions:
    18L×5W×5" H
  • Material:
    Nickel-plated zinc clamp and rod and steel base
  • Cat. No.:
    MSPP-EW0802902

Specifications

About this item

Ultra Flex support system offers the ultimate in flexibility for laboratory use.

  • Flexible arms are extremely versatile and can be placed in virtually any position or angle
  • Designed to fit easily on benchtops or in fume hoods

Each Ultra Flex support system includes three interchangeable clamp heads: two-prong clamp head that opens to 2.6" (6.6 cm) Ø, three-prong clamp head that opens to 2.3" (5.8 cm) Ø, spring clamp head that opens to 0.5" (1.3 cm) Ø and utility wrench.

Ultra Flex Support System with base plate includes flex arm, interchangeable clamp heads, and base plate. Designed to fit easily on benchtops or in fume hoods with a stable 5×5" (12.7×12.7 cm), chemical-resistant black finish steel base plate.