Order Entry
United States
Orders LinkContactUsLinkComponent
4-Acryloylmorpholine 96% stabilised

4-Acryloylmorpholine 96% stabilised

Corrosive
Irritant
Health
Supplier:   Ambeed
4-Acryloylmorpholine 96% stabilised
Supplier:  Ambeed
Certificates
4-Acryloylmorpholine 96% stabilised

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: for Research Use Only

  • CAS Number:5117-12-4
  • Formula:C₇H₁₁NO₂
  • MDL Number:MFCD00047413
  • Molecular Weight:141.17 g/mol
  • Boiling Pt:442.6 °C (760 mm Hg)
  • Melting Pt:35 °C (760 mm Hg)
  • Melting Pt:…35 °C at N/A mmHg
  • Flash Pt:199 °C
  • Storage Temperature:Refrigerator
  • UN:1760
  • ADR:8,Ⅲ
  •  :Stabilised with 4-methoxyphenol (MEHQ)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $88.00
    $88.00
    Each(1/Itm/Bx)
       $88.00

    Supplier Number: A522584-500G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • Glass bottle
    5 g
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $15.00
    $15.00
    Each (25g)
       $15.00

    Supplier Number: A522584-25G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.