ContactUsLinkComponent
Quidel QuickVue RSV Test, NDC
Quidel QuickVue RSV Test, NDC
Catalog # 76474-228
CAS Number:  
undefined
Quidel QuickVue RSV Test, NDC
Catalog # 76474-228
Supplier Number:  QUI 20193
CAS Number:  
Specifications
  • Description:
    QuickVue RSV test
  • Cat. No.:
    76474-228
Specifications
About this item
The QuickVue RSV Test is a dipstick immunoassay that detects respiratory syncytial virus (RSV) antigen (viral fusion protein) directly from nasopharyngeal swab, nasopharyngeal aspirate, or nasal/nasopharyngeal wash specimens for symptomatic pediatric patients (18 years of age and younger).
  • Processing time of 15 minutes or less
  • CLIA complexity is waved