Rapid Response™ Fecal Immunochemical Test (FIT) Control Solutions, BTNX
Catalog # 76473-474
Supplier: BTNX INC.
CAS Number:
Specifications
- Description:IFOBT external controls
- Cat. No.:76473-474
Specifications
About this item
The Rapid Response® Fecal immunochemical test (FIT) positive and negative control solutions are meant to be used with the Rapid Response® FIT Tests.
- Performs approximately 10 tests per level (10 negative tests and 10 positive tests)
- Quantity: 1 pair/kit
- Time to result: 5 minutes
- Storage condition: 15 to 30 °C (59 to 86 °F)
Interpretation of test results are: positive (two lines), negative (one line), invalid (no lines or no control line). The FIT controls are stable at room temperature for the duration of the expiration dating.
Delivery information: Includes 1 positive control vial (0.8 ml), 1 negative control vial (0.8 ml), 4 FIT buffer vials (2 ml each), product insert.