Fraser Broth, Modified, Hardy Diagnostics
Catalog # 76436-462
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Pack type:PC bottle, 500 ml
- Description:Fraser broth, modified
- Size:10/pack
- Cat. No.:76436-462
- Fill volume:225 ml
Specifications
About this item
Fraser Broth, Modified is recommended for the selective enrichment of Listeria spp. in foods and environmental samples.
- Heavy duty bottles are tough and sturdy to reduce breakage
- Easy-to-remove, perforated, shrink wrap ensures product is leak-proof
- Optically clear bottles are made of clarified polycarbonate for clear viewing
- Each bottle is easy to read with a clearly marked measuring fill line