Chix® Quix Plus Cleaning and Sanitizing Towels, Chicopee®
Catalog # 76469-038
Supplier: Janitorial Supplies
Specifications
- Color:Pink
- Material:Cloth
- Cat. No.:76469-038
- Packaging:72/carton
- W×L:13.5×20"
Specifications
About this item
Handy towels are pretreated with a highly concentrated cleaning formula, enabling wet-and-go performance.
- Pretreated with a highly concentrated cleaning formula
- Kills most common germs when used on hard surfaces
- Easy-dispense packaging
Antimicrobial agent kills most common germs in 60 seconds. Easy-dispense packaging. Single-ply towels are ideal for general-purpose cleaning applications.