ContactUsLinkComponent
Traffic Sign, 'VACCINATION STATION AHEAD', Accuform®
Traffic Sign, 'VACCINATION STATION AHEAD', Accuform®
Catalog # 76471-358
undefined
Traffic Sign, 'VACCINATION STATION AHEAD', Accuform®
Catalog # 76471-358
Supplier Number:  FRP803RA
Specifications
  • Description:
    Traffic sign
  • Dimensions:
    12×18"
  • Material:
    Aluminum, reflective
  • Environmentally Preferable:
  • Cat. No.:
    76471-358
  • Legend:
    VACCINATION STATION AHEAD
Specifications
About this item
Traffic signs safely direct and regulate the flow of vehicles and pedestrians in and around parking areas, dock areas, and roadways.
  • Made from durable 0.080" aluminum that will not rust
  • Engineer-grade prismatic reflective sign face provides added visibility in low-light conditions
  • Meets ASTM D 4956 Type I
Message and graphics are printed using UV-resistant inks to prevent fading during long-term outdoor use. Finishing includes clear UV protective overcoat and post mount holes.