yDcpS
4000 units
Catalog # 76339-732
Supplier: New England Biolabs (NEB)
Supplier #: M0463S
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
yDcpS is the scavenging decapping enzyme from S. cerevisiae that hydrolyzes the phosphodiester bond between the gamma and beta phosphates of m7G-capped mRNA, producing diphosphorylated 5 ends and m7GMP. yDcpS is capable of decapping mRNAs of various lengths, decaps Cap0 and Cap1 with equal efficiency, and the diphosphorylated 5 end it produces is suitable for recapping with Vaccinia capping enzyme.
- Provided at high enzyme concentration to facilitate complete decapping
- Active on capped RNA of various sizes (tested on in vitro capped transcripts from 25 nt to 1400 nt)
- Decapped RNA 5' ends can be recapped with Vaccinia capping system supports tagging of mRNA with desthiobiotin-labeled GTP
Specifications
Specifications
- Size:4000 units
- Cat. No.:76339-732
- Concentration:200000 units/ml
- Enzyme Name:yDcpS