ContactUsLinkComponent
Luna Cell Ready Lysis Module, New England Biolabs
Luna Cell Ready Lysis Module, New England Biolabs
Catalog # 76407-360
CAS Number:  
undefined
Luna Cell Ready Lysis Module, New England Biolabs
Catalog # 76407-360
Supplier Number:  E3032S
CAS Number:  
Specifications
  • Description:
    Luna Cell ready lysis module
  • Size:
    100 reactions
  • Cat. No.:
    76407-360
Specifications
About this item
Cell cultures are often analyzed for gene expression or treatment responses as a proxy for a living organism. Traditionally, RNA is extracted and purified from treated cells via column-based or chemical methods. The Luna Cell Ready Lysis Module is a fast, convenient, and sensitive alternative for evaluating RNA expression levels without performing traditional RNA extraction steps.
  • Coordinated cell lysis, RNA release, and genomic DNA removal in a fast 15-minute protocol
  • Effective cell lysis preparation from 10 to 100000 cells across numerous cell lines
  • Minimal sample loss compared to alternative RNA extraction methods, especially at low cell input
  • Compatible with high-throughput applications: cell lysis can take place at room temperature
The module coordinates cell lysis, RNA release, and genomic DNA removal simultaneously, resulting in cell lysate ready for direct RT-qPCR analysis with Luna Universal One-Step RT-qPCR kits. Similar to other Luna products, the lysis buffer includes an inert blue tracking dye for visual assistance throughout the workflow.