Mouse sE-Selectin PicoKine; Quick ELISA Kit, Boster
Catalog # 76467-350
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Pack type:Plastic box
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Target protein:sE-selectin
- Size:96 wells/kit, with removable strips
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76467-350
Specifications
About this item
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Mouse E Selectin/SELE with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum and plasma (heparin, citrate). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. E Selectin/SELE tissue specificity.