Mouse GM-CSF PicoKine; Quick ELISA Kit, Boster
Catalog # 76467-330
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Pack type:Plastic box
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Target protein:GM-CSF
- Size:96 wells/kit, with removable strips
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76467-330
Specifications
About this item
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Mouse Gm-Csf/CSF2 with <1 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Gm-Csf/CSF2 tissue specificity.