Human IL15RA ELISA Kit EZ-Set (DIY Antibody Pairs), Boster
Catalog # 76467-306
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Pack type:Plastic box
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:IL-15RA
- Size:For 5 plates, 96 wells each
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76467-306
Specifications
About this item
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects IL15RA in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.