To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Pack type:Plastic box
- Assay Type:Sandwich
- Host:
- Primary antibody reactivity:Mouse
- Target protein:sE-selectin
- Size:For 5 plates, 96 wells each, reagents only, plates are not included
- Environmentally Preferable:
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76467-176
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects E-Selectin in Mouse samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.