To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:E.coli-derived human FDPS/FPS recombinant protein (Position: S29-Q397).
- Antigen Symbol:FDPS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 ug/vial
- Environmentally Preferable:
- Form:Lyophilized
- Storage Temperature:At -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquoted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
- Immunogen:E.coli-derived human FDPS/FPS recombinant protein (Position: S29-Q397).
- Purification:Immunogen affinity purified.
- Cat. No.:76464-618
Rabbit IgG polyclonal antibody for FDPS/FPS detection. Tested with WB, FCM, Direct ELISA in Human,Rat.
Type: Primary
Antigen: FDPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Rat