About this item
The Exo-CIP Rapid PCR cleanup kit contains optimized formulations of thermolabile Exonuclease I and thermolabile Calf Intestinal Phosphatase (CIP) and is used to rapidly degrade residual PCR primers and dephosphorylate excess dNTPs after amplification.
- Rapidly degrade residual PCR primers and dephosphorylate excess dNTPs after amplification
- Reaction complete in 4 minutes
- Thermolabile formulation can be heat inactivated in 1 minute at 80 °C
- PCR product can be used directly in downstream applications
Degradation occurs in only 4 minutes at 37 °C, and is immediately followed by rapid inactivation of the enzymes by heating for 1 minute at 80 °C. In just 5 minutes, the PCR product is ready for downstream analysis, such as Sanger Sequencing, SNP detection, or library preparation for NGS. The Exo-CIP Rapid PCR cleanup kit is compatible with all commonly-used PCR buffers.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: