ContactUsLinkComponent
SHuffle T7 Competent E. coli, New England Biolabs
SHuffle T7 Competent E. coli, New England Biolabs
  103391-802
undefined
SHuffle T7 Competent E. coli, New England Biolabs
  103391-802
 :  C3026J
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    SHuffle T7 competent E.coli
  • Size:
    12 vials×0.05 ml/tube
  • Cat. No.:
    103391-802
Chemically competent E. coli K12 cells suitable for T7 protein expression with enhanced capacity to correctly fold proteins with multiple disulfide bonds in the cytoplasm.
  • T7 expression
  • Engineered E. coli K12 to promote disulfide bond formation in the cytoplasm
  • Resistant to phage T1 (fhuA2)Free of animal products
  • NEB does not add a dry ice surcharge to our competent cell shipments